Merck
  • L-kynurenine/aryl hydrocarbon receptor pathway mediates brain damage after experimental stroke.
L-kynurenine/aryl hydrocarbon receptor pathway mediates brain damage after experimental stroke.
Circulation (2014-11-02)
María I Cuartero, Iván Ballesteros, Juan de la Parra, Andrew L Harkin, Aine Abautret-Daly, Eoin Sherwin, Pedro Fernández-Salguero, Angel L Corbí, Ignacio Lizasoain, María A Moro
ABSTRACT
Aryl hydrocarbon receptor (AhR) is a transcription factor that belongs to the basic helix-loop-helix PAS (Per-Arnt-Sim homology domain) family known to mediate the toxic and carcinogenic effects of xenobiotics. Interestingly, AhR is widely expressed in the central nervous system, but its physiological and pathological roles are still unclear. To define the role of AhR in stroke, we used middle cerebral artery occlusion in mice and oxygen-glucose deprivation in rat cortical neurons. The results presented here show that the ischemic insult increases total and nuclear AhR levels and AhR transcriptional activity in neurons in vivo and in vitro. We also show that AhR has a causal role in acute ischemic damage because pharmacological or genetic loss-of-function approaches result in neuroprotection. Inhibition of cAMP response element-binding protein-dependent signaling may participate in the deleterious actions of AhR. Finally, we have also found that L-kynurenine, a tryptophan metabolite with AhR agonistic properties, is an endogenous ligand that mediates AhR activation in the brain after middle cerebral artery occlusion. Our data demonstrate that an L-kynurenine/AhR pathway mediates acute brain damage after stroke and open new possibilities for the diagnosis and treatment of this pathology.
Product Description
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
680C91, ≥98% (HPLC)
Sigma-Aldrich
6, 2′, 4′-trimethoxyflavone, ≥98% (HPLC)
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Tryptophan, European Pharmacopoeia (EP) Reference Standard