Merck
  • Prediction of serine/threonine phosphorylation sites in bacteria proteins.
Prediction of serine/threonine phosphorylation sites in bacteria proteins.
Advances in experimental medicine and biology (2014-11-13)
Zhengpeng Li, Ping Wu, Yuanyuan Zhao, Zexian Liu, Wei Zhao
ABSTRACT
As a critical post-translational modification, phosphorylation plays important roles in regulating various biological processes, while recent studies suggest that phosphorylation in bacteria is also critical for functional signaling transduction. Since identification of phosphorylation substrates and sites is fundamental for understanding the phosphorylation mediated regulatory mechanism, a number of studies have been contributed to this area. Since experimental identification of phosphorylation sites is time-consuming and labor-intensive, computational predictions attract much attention for its convenience to provide helpful information. However, although there are a large number of computational studies in eukaryotes, predictions in bacteria are still rare. In this study, we present a new predictor of cPhosBac to predict phosphorylation serine/threonine in bacteria proteins. The predictor is developed with CKSAAP algorithm, which was combined with motif length selection to optimize the prediction, which achieves promising performance. The online service of cPhosBac is available at: http://netalign.ustc.edu.cn/cphosbac/ .
Product Description
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Serine, European Pharmacopoeia (EP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Threonine
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland