Merck
  • Glycosuria-mediated urinary uric acid excretion in patients with uncomplicated type 1 diabetes mellitus.
Glycosuria-mediated urinary uric acid excretion in patients with uncomplicated type 1 diabetes mellitus.
American journal of physiology. Renal physiology (2014-11-08)
Yuliya Lytvyn, Marko Škrtić, Gary K Yang, Paul M Yip, Bruce A Perkins, David Z I Cherney
ABSTRACT
Plasma uric acid (PUA) is associated with metabolic, cardiovascular, and renal abnormalities in patients with type 2 diabetes but is less well understood in type 1 diabetes (T1D). Our aim was to compare PUA levels and fractional uric acid excretion (FEUA) in patients with T1D vs. healthy controls (HC) during euglycemia and hyperglycemia. PUA, FEUA, blood pressure (BP), glomerular filtration rate (GFR-inulin), and effective renal plasma flow (ERPF-paraaminohippurate) were evaluated in patients with T1D (n = 66) during clamped euglycemia (glucose 4-6 mmol/l) and hyperglycemia (9-11 mmol/l), and in HC (n = 41) during euglycemia. To separate the effects of hyperglycemia vs. increased glycosuria, parameters were evaluated during clamped euglycemia in a subset of T1D patients before and after sodium glucose cotransporter 2 (SGLT2) inhibition for 8 wk. PUA was lower in T1D vs. HC (228 ± 62 vs. 305 ± 75 μmol/l, P < 0.0001). In T1D, hyperglycemia further decreased PUA (228 ± 62 to 199 ± 65 μmol/l, P < 0.0001), which was accompanied by an increase in FEUA (7.3 ± 3.8 to 11.6 ± 6.7, P < 0.0001). In T1D, PUA levels correlated positively with SBP (P = 0.029) and negatively with ERPF (P = 0.031) and GFR (P = 0.028). After induction of glycosuria with SGLT2 inhibition while maintaining clamped euglycemia, PUA decreased (P < 0.0001) and FEUA increased (P < 0.0001). PUA is lower in T1D vs. HC and positively correlates with SBP and negatively with GFR and ERPF in T1D. Glycosuria rather than hyperglycemia increases uricosuria in T1D. Future studies examining the effect of uric acid-lowering therapies should account for the impact of ambient glycemia, which causes an important uricosuric effect.
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline