Merck
  • Assessment of DNA synthesis in Islet-1⁺ cells in the adult murine heart.
Assessment of DNA synthesis in Islet-1⁺ cells in the adult murine heart.
Biochemical and biophysical research communications (2014-12-03)
Florian Weinberger, Dennis Mehrkens, Jutta Starbatty, Philipp Nicol, Thomas Eschenhagen
ABSTRACT
Islet-1 positive (Islet-1(+)) cardiac progenitor cells give rise to the right ventricle, atria and outflow tract during murine cardiac development. In the adult heart Islet-1 expression is limited to parasympathetic neurons, few cardiomyocytes, smooth muscle cells, within the proximal aorta and pulmonary artery and sinoatrial node cells. Its role in these cells is unknown. Here we tested the hypothesis that Islet-1(+) cells retain proliferative activity and may therefore play a role in regenerating specialized regions in the heart. DNA synthesis was analyzed by the incorporation of tritiated thymidine ((3)H-thymidine) in Isl-1-nLacZ mice, a transgenic model with an insertion of a nuclear beta-galactosidase in the Islet-1 locus. Mice received daily injections of (3)H-thymidine for 5 days. DNA synthesis was visualized throughout the heart by dipping autoradiography of cryosections. Colocalization of an nLacZ-signal and silver grains would indicate DNA synthesis in Islet-1(+) cells. Whereas Islet(-) non-myocyte nuclei were regularly marked by accumulation of silver grains, colocalization with nLacZ-signals was not detected in >25,000 cells analyzed. Islet-1(+) cells are quiescent in the adult heart, suggesting that, under normal conditions, even pacemaking cells do not proliferate at higher rates than normal cardiac myocytes.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)