Merck
  • Enhanced anticancer activity of a protein phosphatase 2A inhibitor on chemotherapy and radiation in head and neck squamous cell carcinoma.
Enhanced anticancer activity of a protein phosphatase 2A inhibitor on chemotherapy and radiation in head and neck squamous cell carcinoma.
Cancer letters (2014-12-03)
Dong-wang Zhu, Yong-xiang Yuan, Jin-ke Qiao, Cong Yu, Xi Yang, Li-zhen Wang, Zhi-yuan Zhang, Lai-ping Zhong
ABSTRACT
The aim of this study is to eliminate more cancer cells by promoting them from quiescence into cell cycle or by changing their molecular events, leading them to be sensitive to radiation or chemotherapy. Protein phosphatase 2A plays an important role in many cellular functions and regulates various biological processes. It is unclear that LB1, which is an inhibitor of protein phosphatase 2A, has enhanced anticancer activity on chemotherapy (cisplatin and 5-fluorourcil) and radiation in head and neck squamous cell carcinoma (HNSCC). Herein, we performed both in vitro and in vivo studies to determine the anticancer activity of LB1 on chemotherapy and radiation in HNSCC, with detection of p53 expression, AKT and MDM2 phosphorylation. In vitro studies indicated that, LB1 could significantly enhance the cytotoxicity of cisplatin, 5-fluorourcil, and radiation; LB1 could also significantly enhance the treatment effect of cisplatin in nude mice. The anticancer activity of LB1 was mediated by increased AKT phosphorylation and decreased p53 expression with increased MDM2 phosphorylation, especially when combined with cisplatin. Our data suggest a strategy of improving treatment effect through the enhanced anticancer activity of LB1 on cisplatin-based chemotherapy and radiation in HNSCC.
Product Description
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Serine, European Pharmacopoeia (EP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard