Merck
  • Associations of cadmium and lead exposure with leukocyte telomere length: findings from National Health and Nutrition Examination Survey, 1999-2002.
Associations of cadmium and lead exposure with leukocyte telomere length: findings from National Health and Nutrition Examination Survey, 1999-2002.
American journal of epidemiology (2014-12-17)
Ami R Zota, Belinda L Needham, Elizabeth H Blackburn, Jue Lin, Sung Kyun Park, David H Rehkopf, Elissa S Epel
ABSTRACT
Cadmium and lead are ubiquitous environmental contaminants that might increase risks of cardiovascular disease and other aging-related diseases, but their relationships with leukocyte telomere length (LTL), a marker of cellular aging, are poorly understood. In experimental studies, they have been shown to induce telomere shortening, but no epidemiologic study to date has examined their associations with LTL in the general population. We examined associations of blood lead and cadmium (n = 6,796) and urine cadmium (n = 2,093) levels with LTL among a nationally representative sample of US adults from the National Health and Nutrition Examination Survey (1999-2002). The study population geometric mean concentrations were 1.67 µg/dL (95% confidence interval (CI): 1.63, 1.70) for blood lead, 0.44 µg/L (95% CI: 0.42, 0.47) for blood cadmium, and 0.28 µg/L (95% CI: 0.27, 0.30) for urine cadmium. After adjustment for potential confounders, the highest (versus lowest) quartiles of blood and urine cadmium were associated with -5.54% (95% CI: -8.70, -2.37) and -4.50% (95% CI: -8.79, -0.20) shorter LTLs, respectively, with evidence of dose-response relationship (P for trend < 0.05). There was no association between blood lead concentration and LTL. These findings provide further evidence of physiological impacts of cadmium at environmental levels and might provide insight into biological pathways underlying cadmium toxicity and chronic disease risks.
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Lead, wire reel, 2m, diameter 3.0mm, as drawn, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Lead, wire reel, 5m, diameter 0.5mm, as drawn, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 6mm disks, thickness 0.01mm, 99.7+%
Cadmium, tube, 1000mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.7+%
Lead, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%