Merck
  • Maternal folate status, but not that of vitamins B-12 or B-6, is associated with gestational age and preterm birth risk in a multiethnic Asian population.
Maternal folate status, but not that of vitamins B-12 or B-6, is associated with gestational age and preterm birth risk in a multiethnic Asian population.
The Journal of nutrition (2014-12-21)
Ling-Wei Chen, Ai Lin Lim, Marjorelee Colega, Mya-Thway Tint, Izzuddin M Aris, Chuen Seng Tan, Yap-Seng Chong, Peter D Gluckman, Keith M Godfrey, Kenneth Kwek, Seang-Mei Saw, Fabian Yap, Yung Seng Lee, Mary Foong-Fong Chong, Rob M van Dam
ABSTRACT
Maternal folate, vitamin B-12, and vitamin B-6 concentrations during pregnancy have been shown to influence birth outcomes, but the evidence is inconclusive. We aimed to examine the associations of maternal B-vitamin status with gestational age, birth weight, and length in a birth cohort study in Singapore. Maternal blood samples (n = 999) collected during weeks 26-28 of gestation were assayed for plasma folate, vitamin B-12, and vitamin B-6 concentrations. Birth weight and gestational age data were obtained from hospital records, and other anthropometric variables were measured within 72 h after birth. Relations between B-vitamin status and birth outcomes were assessed by linear or logistic regression with adjustment for potential confounders. Median (IQR) plasma concentrations were 34.4 (24.5-44.6) nmol/L for folate, 209 (167-258) pmol/L for vitamin B-12, and 61.8 (25.9-113) nmol/L for vitamin B-6. We found that higher plasma folate concentrations were associated with a longer gestational age (0.12 wk per SD increase in folate; 95% CI: 0.02, 0.21) and tended to be associated with lower risk of all preterm birth (delivery at <37 wk of gestation; OR: 0.79; 95% CI: 0.63, 1.00) and spontaneous preterm birth (OR: 0.76; 95% CI: 0.56, 1.04). Overall, concentrations of maternal folate, vitamin B-12, and vitamin B-6 were not independently associated with birth weight or being born small for gestational age (SGA; birth weight <10th percentile for gestational age). Higher maternal folate concentrations during late pregnancy were associated with longer gestational age and tended to be associated with a lower risk of preterm birth in this multiethnic Asian population. In contrast, the results of our study suggested little or no benefit of higher folate concentrations for reducing the risk of SGA or of higher vitamin B-6 and vitamin B-12 concentrations for reducing the risk of preterm birth or SGA.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Folic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, meets USP testing specifications