Merck
  • Stable gold(III) catalysts by oxidative addition of a carbon-carbon bond.
Stable gold(III) catalysts by oxidative addition of a carbon-carbon bond.
Nature (2015-01-23)
Chung-Yeh Wu, Takahiro Horibe, Christian Borch Jacobsen, F Dean Toste
ABSTRACT
Low-valent late transition-metal catalysis has become indispensable to chemical synthesis, but homogeneous high-valent transition-metal catalysis is underdeveloped, mainly owing to the reactivity of high-valent transition-metal complexes and the challenges associated with synthesizing them. Here we report a carbon-carbon bond cleavage at ambient conditions by a Au(i) complex that generates a stable Au(iii) cationic complex. In contrast to the well-established soft and carbophilic Au(i) catalyst, this Au(iii) complex exhibits hard, oxophilic Lewis acidity. For example, we observed catalytic activation of α,β-unsaturated aldehydes towards selective conjugate additions as well as activation of an unsaturated aldehyde-allene for a [2 + 2] cycloaddition reaction. The origin of the regioselectivity and catalytic activity was elucidated by X-ray crystallographic analysis of an isolated Au(iii)-activated cinnamaldehyde intermediate. The concepts revealed suggest a strategy for accessing high-valent transition-metal catalysis from readily available precursors.
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Gold coated glass cover slip, layer thickness 100 Å, 99.999% (Au), L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Carbon, rod, 100mm, diameter 13.0mm, graphite, 100%
Carbon, rod, 25mm, diameter 50mm, graphite, 99.95%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg