Merck
  • Nox4 is a major source of superoxide production in human brain pericytes.
Nox4 is a major source of superoxide production in human brain pericytes.
Journal of vascular research (2015-01-24)
Junya Kuroda, Tetsuro Ago, Ataru Nishimura, Kuniyuki Nakamura, Ryu Matsuo, Yoshinobu Wakisaka, Masahiro Kamouchi, Takanari Kitazono
ABSTRACT
Pericytes are multifunctional cells surrounding capillaries and postcapillary venules. In brain microvasculature, pericytes play a pivotal role under physiological and pathological conditions by producing reactive oxygen species (ROS). The aims of this study were to elucidate the source of ROS and its regulation in human brain pericytes. The expression of Nox enzymes in the cells was evaluated using RT-PCR and western blot. Superoxide production was determined by superoxide dismutase-inhibitable chemiluminescence. Silencing of Nox4 was performed using RNAi, and cell proliferation was evaluated using the 3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium (MTS) assay. Nox4 was predominant among the Nox family in human brain pericytes. Membrane fractions of cells produced superoxide in the presence of NAD(P)H. Superoxide production was almost abolished with diphenileneiodonium, a Nox inhibitor; however, inhibitors of other possible superoxide-producing enzymes had no effect on NAD(P)H-dependent superoxide production. Pericytes expressed angiotensin II (Ang II) receptors, and Ang II upregulated Nox4 expression. Hypoxic conditions also increased the Nox4 expression. Silencing of Nox4 significantly reduced ROS production and attenuated cell proliferation. Our study showed that Nox4 is a major superoxide-producing enzyme and that its expression is regulated by Ang II and hypoxic stress in human brain pericytes. In addition, Nox4 may promote cell growth.
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder