Merck
  • Prevalence of thalassaemia, iron-deficiency anaemia and glucose-6-phosphate dehydrogenase deficiency among Arab migrating nomad children, southern Islamic Republic of Iran.
Prevalence of thalassaemia, iron-deficiency anaemia and glucose-6-phosphate dehydrogenase deficiency among Arab migrating nomad children, southern Islamic Republic of Iran.
Eastern Mediterranean health journal = La revue de sante de la Mediterranee orientale = al-Majallah al-sihhiyah li-sharq al-mutawassit (2015-01-21)
M Pasalar, D Mehrabani, A Afrasiabi, Z Mehravar, I Reyhani, R Hamidi, M Karimi
ABSTRACT
This study investigated the prevalence of iron-deficiency anaemia, glucose-6-phosphate dehydrogenase (G6PD) deficiency and β-thalassaemia trait among Arab migrating nomad children in southern Islamic Republic of Iran. Blood samples were analysed from 134 schoolchildren aged < 18 years (51 males, 83 females). Low serum ferritin (< 12 ng/dL) was present in 17.9% of children (21.7% in females and 11.8% in males). Low haemoglobin (Hb) correlated significantly with a low serum ferritin. Only 1 child had G6PD deficiency. A total of 9.7% of children had HbA2 ≥ 3.5 g/dL, indicating β-thalassaemia trait (10.8% in females and 7.8% in males). Mean serum iron, serum ferritin and total iron binding capacity were similar in males and females. Serum ferritin index was as accurate as Hb index in the diagnosis of iron-deficiency anaemia. A high prevalence of β-thalassaemia trait was the major potential risk factor in this population.
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Iron, foil, 4mm disks, thickness 1.0mm, as rolled, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.01mm, 99.99+%
Iron, foil, 2m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, 50m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 1m coil, thickness 0.125mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.005mm, 99.85%
Iron, foil, 15mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.0125mm, 99.85%
Iron, foil, 25mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.015mm, 99.85%
Iron, foil, 10mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.007mm, 99.85%
Iron, foil, 25mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 15mm disks, thickness 0.004mm, 99.85%
Iron, foil, 15mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 10mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 4mm disks, thickness 0.004mm, 99.85%
Iron, foil, 10mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.006mm, 99.85%
Iron, foil, 15mm disks, thickness 0.075mm, hard, 99.5%