Merck
  • Propylphosphonic anhydride (T3P?): an efficient reagent for the one-pot synthesis of 1, 2, 4-oxadiazoles, 1, 3, 4-oxadiazoles, and 1, 3, 4-thiadiazoles.
Propylphosphonic anhydride (T3P?): an efficient reagent for the one-pot synthesis of 1, 2, 4-oxadiazoles, 1, 3, 4-oxadiazoles, and 1, 3, 4-thiadiazoles.
Tetrahedron (2009)
Augustine JK, et al.