Merck
  • A novel mutation in C5L2 gene was associated with hyperlipidemia and retinitis pigmentosa in a Chinese family.
A novel mutation in C5L2 gene was associated with hyperlipidemia and retinitis pigmentosa in a Chinese family.
Lipids in health and disease (2014-06-03)
Ling-Hui Qu, Xin Jin, Liang-Mao Li, Shi-Ying Li, Han-Ping Xie
ABSTRACT
Previous studies indicated that hyperlipidemia was associated with retinitis pigmentosa (RP). We aimed to identify the mutations in the C5L2 gene which was reported to be associated with hyperlipidemia in a Chinese family with (RP). The Proband from the family was screened for mutations in the C5L2 gene that was known to cause hyperlipidemia. Cosegregation analysis was performed in the available family members. Linkage analysis was performed for one missense mutation to calculate the likelihood of its pathogenicity. One hundred and fifty unrelated, healthy Chinese subjects were screened to exclude nonpathogenic polymorphisms. By direct sequencing method, we identified a novel mutation (Thr196Asn) in C5L2 gene. In this family, each affected family members with RP showed a heterozygous mutation in the C5L2 gene. And all the carriers with heterozygous mutation have increased serum lipid levels in this family. The present study has extended the mutation spectrum of C5L2, and Thr196Asn mutations in C5L2 were associated with RP and serum lipid levels.