Merck
  • Pretargeted immunoPET of prostate cancer with an anti-TROP-2 x anti-HSG bispecific antibody in mice with PC3 xenografts.
Pretargeted immunoPET of prostate cancer with an anti-TROP-2 x anti-HSG bispecific antibody in mice with PC3 xenografts.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-07-26)
Catharina M van Rij, Cathelijne Frielink, David M Goldenberg, Robert M Sharkey, Gerben M Franssen, Susanne Lütje, William J McBride, Wim J G Oyen, Otto C Boerman
ABSTRACT
Pretargeting with bispecific antibodies and radiolabeled hapten-peptides could be used to specifically target tumors with high target-to-background ratios. TF12 is a trivalent bispecific antibody that consists of two anti-TROP-2 Fab fragments and one anti-HSG (histamine-succinyl-glycine) Fab fragment. The TROP-2 antigen is expressed in many epithelial cancers, including prostate cancer (PC), and therefore, this bispecific antibody can be used for pretargeting of PC. In this study, the potential for pretargeted radioimmunoPET with TF12 and the (68)Ga-labeled di-HSG peptide IMP288 in mice with human PC xenografts was investigated using 2-deoxy-2-[(18)F]fluoro-D-glucose ([(18)F]FDG) as a reference. The potential of pretargeted immunoPET with TF12 and the (68)Ga-labeled di-HSG hapten-peptide, IMP288, was studied in mice with subcutaneous PC3 tumors using [(18)F]FDG as a reference. Furthermore, the use of this pretargeting system for imaging PC lesions was evaluated in mice with intraperitoneally growing tumors with [(18)F]FDG as a reference. [(68)Ga]lMP288 showed rapid accumulation in the TF12 pretargeted subcutaneous tumor (7.2 ± 1.1 % ID/g) with low uptake in the kidneys (1.8 ± 0.5 % ID/g) and high tumor-to-blood ratios (17.4 ± 11.2) at 1 h p.i. Accumulation of [(18)F]FDG in the s.c. tumors was significantly lower (3.4 ± 0.9 % ID/g, P = 0.008), with lower tumor-to-blood ratios (3.0 ± 1.9, P = 0.011). ImmunoPET/CT images clearly visualized both subcutaneous and intraperitoneal tumors as small as 5 mm(3) with low blood levels and kidney uptake as early as 1 h p.i. Pretargeted immunoPET with TF12 in combination with a (68)Ga-labeled hapten-peptide is an efficient system for rapid, sensitive, and specific imaging of prostate cancer.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC