Merck
  • A retrospective examination of mean relative telomere length in the Tasmanian Familial Hematological Malignancies Study.
A retrospective examination of mean relative telomere length in the Tasmanian Familial Hematological Malignancies Study.
Oncology reports (2014-10-30)
Nicholas B Blackburn, Jac C Charlesworth, James R Marthick, Elizabeth M Tegg, Katherine A Marsden, Velandai Srikanth, John Blangero, Ray M Lowenthal, Simon J Foote, Joanne L Dickinson
ABSTRACT
Telomere length has a biological link to cancer, with excessive telomere shortening leading to genetic instability and resultant malignant transformation. Telomere length is heritable and genetic variants determining telomere length have been identified. Telomere biology has been implicated in the development of hematological malignancies (HMs), therefore, closer examination of telomere length in HMs may provide further insight into genetic etiology of disease development and support for telomere length as a prognostic factor in HMs. We retrospectively examined mean relative telomere length in the Tasmanian Familial Hematological Malignancies Study using a quantitative PCR method on genomic DNA from peripheral blood samples. Fifty-five familial HM cases, 191 unaffected relatives of familial HM cases and 75 non-familial HM cases were compared with 758 population controls. Variance components modeling was employed to identify factors influencing variation in telomere length. Overall, HM cases had shorter mean relative telomere length (p=2.9×10-6) and this was observed across both familial and non-familial HM cases (p=2.2x10-4 and 2.2x10-5, respectively) as well as additional subgroupings of HM cases according to broad subtypes. Mean relative telomere length was also significantly heritable (62.6%; p=4.7x10-5) in the HM families in the present study. We present new evidence of significantly shorter mean relative telomere length in both familial and non-familial HM cases from the same population adding further support to the potential use of telomere length as a prognostic factor in HMs. Whether telomere shortening is the cause of or the result of HMs is yet to be determined, but as telomere length was found to be highly heritable in our HM families this suggests that genetics driving the variation in telomere length is related to HM disease risk.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O