Merck
  • Fructose restores susceptibility of multidrug-resistant Edwardsiella tarda to kanamycin.
Fructose restores susceptibility of multidrug-resistant Edwardsiella tarda to kanamycin.
Journal of proteome research (2015-02-13)
Yu-bin Su, Bo Peng, Yi Han, Hui Li, Xuan-xian Peng
ABSTRACT
Edwardsiella tarda, the causative agent of Edwardsiellosis, imposes medical challenges in both the clinic and aquaculture. The emergence of multidrug resistant strains makes antibiotic treatment impractical. The identification of molecules that facilitate or promote antibiotic efficacy is in high demand. In the present study, we aimed to identify small molecules whose abundance is correlated with kanamycin resistance in E. tarda by gas chromatography-mass spectrometry. We found that the abundance of fructose was greatly suppressed in kanamycin-resistant strains. The incubation of kanamycin-resistant bacteria with exogenous fructose sensitized the bacteria to kanamycin. Moreover, the fructose also functioned in bacteria persisters and biofilm. The synergistic effects of fructose and kanamycin were validated in a mouse model. Furthermore, the mechanism relies on fructose in activating TCA cycle to produce NADH, which generates proton motive force to increase the uptake of the antibiotics. Therefore, we present a novel approach in fighting against multidrug resistant bacteria through exploration of antibiotic-suppressed molecules.
Product Description
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic anhydride, 99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
Adonitol, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%