Merck
  • Antihormonal potential of selected D-homo and D-seco estratriene derivatives.
Antihormonal potential of selected D-homo and D-seco estratriene derivatives.
Steroids (2014-09-11)
Suzana S Jovanović-Šanta, Edward T Petri, Olivera R Klisurić, Mihály Szécsi, Radmila Kovačević, Julijana A Petrović
ABSTRACT
Since many estrogen derivatives exhibit anti-hormone or enzyme inhibition potential, a large number of steroidal derivatives have been synthesised from appropriate precursors, in order to obtain potential therapeutics for the treatment of hormone-dependent cancers. In molecular docking studies, based on X-ray crystallographic analysis, selected D-homo and D-seco estratriene derivatives were predicted to bind strongly to estrogen receptor α (ERα), aromatase and 17,20 lyase, suggesting they could be good starting compounds for antihormonal studies. Test results in vivo suggest that these compounds do not possess estrogenic activity, while some of them showed weak anti-estrogenic properties. In vitro anti-aromatase and anti-lyase assays showed partial inhibition of these two enzymes, while some compounds activated aromatase. Aromatase activators are capable of promoting estrogen synthesis for treatment of pathological conditions caused by estrogen depletion, e.g. osteopenia or osteoporosis.
Product Description
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Aminoglutethimide
Sigma-Aldrich
Estradiol, meets USP testing specifications