Merck
  • Efficient visible-light-driven photocatalytic degradation of nitrophenol by using graphene-encapsulated TiO₂ nanowires.
Efficient visible-light-driven photocatalytic degradation of nitrophenol by using graphene-encapsulated TiO₂ nanowires.
Journal of hazardous materials (2014-10-14)
Hyun-Gyu Lee, Gopalan Sai-Anand, Shanmugasundaram Komathi, Anantha-Iyengar Gopalan, Shin-Won Kang, Kwang-Pill Lee
ABSTRACT
In this work, a new hybrid nanocatalyst, namely titanium dioxide (TiO2) composite nanowires, encapsulated with graphene (G) and palladium nanoparticles (Pd NPs) (designated as G-Pd@TiO2-CNWs), was prepared. In preparing the nanowires, a combination of electrospinning and hydrothermal approaches was employed. The visible-light-driven photocatalytic performance of G-Pd@TiO2-CNWs was investigated using the reduction of 4-nitrophenol (4-NP) as a model reaction. The results showed that G-Pd@TiO2-CNWs converted nearly 100% of 4-NP under visible light irradiation. The reaction kinetics of the photocatalytic reduction of 4-NP was studied by UV-vis spectrophotometry and the apparent rate constant was determined and compared with those for other supported TiO2 catalysts. Furthermore, the spent G-Pd@TiO2-CNWs could be recovered by simple centrifugation and reused. The work is expected to shed new light on the development of G-incorporated hybrid nanostructures for harvesting light energy and on the development of new photocatalysts for the removal of environmental pollutants.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
VenPure® SF, powder
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%