Merck
  • Aqueous two-phase extraction for determination of triazine herbicides in milk by high-performance liquid chromatography.
Aqueous two-phase extraction for determination of triazine herbicides in milk by high-performance liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Xiao Yang, Rui Yu, Shaohua Zhang, Bocheng Cao, Zhongling Liu, Lei Lei, Na Li, Zhibing Wang, Liyuan Zhang, Hanqi Zhang, Yanhua Chen
ABSTRACT
A simple extraction method based on acetonitrile-K2HPO4 aqueous two-phase system was developed for separation and enrichment of five triazines in milk samples. Acetonitrile was used for extraction of analytes from milk sample and precipitation of milk protein. Deproteinization and extraction were achieved in one single step. Analytes were extracted into the upper phase of the aqueous two-phase system. The parameters affecting the extraction efficiency, such as the volume of acetonitrile, the type and amount of salts, pH value of sample and extraction time were investigated. The limits of detection of atraton, desmetryn, atrazine, terbumeton and terbuthylazine were 2.1, 2.6, 2.3, 2.8 and 2.5μg/L, respectively. When the present method was applied to the analysis of real milk samples, the recoveries of analytes ranged from 86.3 to 120.6% and relative standard deviations were lower than 7.9%.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%