Merck
  • Determination and pharmacokinetic studies of arecoline in dog plasma by liquid chromatography-tandem mass spectrometry.
Determination and pharmacokinetic studies of arecoline in dog plasma by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-21)
Bing Li, Xu-Zheng Zhou, Jian-Yong Li, Ya-Jun Yang, Jian-Rong Niu, Xiao-Juan Wei, Xi-Wang Liu, Jin-Shan Li, Ji-Yu Zhang
ABSTRACT
A rapid and sensitive high-performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the determination of arecoline concentration in dog plasma. Plasma sample was prepared by protein precipitation using n-hexane (containing 1% isoamyl alcohol) with β-pinene as an internal standard. Chromatographic separation was achieved on an Agilent C18 column (4.6×75mm, 3.5μm) using methanol: 5mM ammonium acetate as the mobile phase with isocratic elution. Mass detection was carried out using positive electrospray ionization in multiple reaction monitoring mode. The calibration curve for arecoline was linear over a concentration range of 2-500ng/mL. The intra-day and inter-day accuracy and precision were within the acceptable limits of ±10% at all concentrations. In summary, the LC-MS/MS method described herein was fully validated and successfully applied to the pharmacokinetic study of arecoline hydrobromide tablets in dogs after oral administration.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Arecoline hydrobromide, VETRANAL®, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard