Merck
  • Primary and secondary metabolite profiling of doenjang, a fermented soybean paste during industrial processing.
Primary and secondary metabolite profiling of doenjang, a fermented soybean paste during industrial processing.
Food chemistry (2014-07-21)
Su Yun Lee, Sarah Lee, Sunmin Lee, Ji Young Oh, Eun Jung Jeon, Hyung Seok Ryu, Choong Hwan Lee
ABSTRACT
In this study, a comprehensive metabolite profile analysis of doenjang, a fermented soybean paste, at various steps of its industrial 5-step production process was conducted, by combining gas and liquid chromatography-mass spectrometry techniques with multivariate analysis. From the partial least squares discriminant analysis of primary and secondary metabolites, the patterns were clearly distinguishable between the various processing steps (step 1: steaming, step 2: drying, step 3: meju fermentation, step 4: brining, step 5: doenjang aging). Of the primary metabolites, most of the monosaccharides, amino acids, and fatty acids increased in steps 3-5. Isoflavone and soyasaponin derivatives were major secondary metabolites identified during the processing of doenjang. Isoflavone glycosides gradually decreased after step 1, while isoflavone aglycones distinctly increased in steps 4-5. Soyasaponins generally decreased during processing after step 2. Increased isoflavone aglycones, such as daidzein, glycitein, and genistein, were observed in steps 4-5 showed the strongest positive correlation with doenjang's antioxidant potential and total phenolic content.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)