Merck
Development of a nanoemulsion of Phyllanthus emblica L. branch extract.
Drug development and industrial pharmacy (2013-09-24)
Rungsiri Chaiittianan, Bungorn Sripanidkulchai
ABSTRACT
For potential topical administration, we formulated a nanoemulsion containing phenolic constituents of Phyllanthus emblica branch extract. The nanoemulsion has high entrapment efficiency, small particle size, is stable, and can release its main chemical components. Branches of P. emblica were extracted with 50% ethanol (EPE) with 5.4% yield. HPLC analysis indicated several phenolic compounds, including gallic acid, vanillic acid, epigallocatechin (EGC), epigallocatechin gallate (EGCG) and ellagic acid. These were selected as chemical markers of EPE in the nanoemulsion development. The nanoemulsion was prepared by microemulsion techniques with hot high pressure homogenization. A ternary phase diagram was constructed to obtain the optimized nanoemulsion. The obtained transparent EPE nanoemulsion is composed of isopropyl myristate (0.6% w/w), Brij® 78 (0.35% w/w), and 0.15% (w/w) EPE. The optimized EPE nanoemulsion had a median particle size of 191.63 ± 4.07 nm with a narrow particle size distribution, a zeta potential of -10.19 ± 0.54 mV, high entrapment efficiency at 67.99 ± 0.87% and good stability at 4 °C after 90 d of storage. The release of active ingredients from the EPE nanoemulsion was slower than that of the EPE aqueous formulation. The loading ratios of the five phenolic compounds were high, with relative order of EGC > EGCG > vanillic acid > gallic acid > ellagic acid, resulting in slow release profiles of EGC and EGCG in the EPE nanoemulsion. In conclusion, the obtained EPE nanoemulsion has good characteristics for future clinical trials.
Product Description
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Epigallocatechin gallate, primary reference standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)