Merck
  • Oxidative degradation of organic pollutants in aqueous solution using zero valent copper under aerobic atmosphere condition.
Oxidative degradation of organic pollutants in aqueous solution using zero valent copper under aerobic atmosphere condition.
Journal of hazardous materials (2014-05-27)
Gang Wen, Sheng-Jun Wang, Jun Ma, Ting-Lin Huang, Zheng-Qian Liu, Lei Zhao, Jin-Lan Xu
ABSTRACT
Oxidative degradation of organic pollutants and its mechanism were investigated in aqueous solution using zero valent copper (ZVC) under aerobic atmosphere condition. Diethyl phthalate (DEP) was completely oxidized after 120 min reaction by ZVC at initial pH 2.5 open to the air. DEP degradation followed the pseudo-first-order kinetics after the lag period, and the degradation rate of DEP increased gradually with the increase of ZVC dosage, and the decrease of initial pH from 5.8 to 2.0. ZVC required a shorter induction time and exhibited persistent oxidation capacity compared to that of zero valent iron and zero valent aluminium. The mechanism investigation showed that remarkable amount of Cu(+)/Cu(2+) and H2O2 were formed in ZVC acidic system, which was due to the corrosive dissolution of ZVC and the concurrent reduction of oxygen. The addition of tert-butanol completely inhibited the degradation of DEP and the addition of Fe(2+) greatly enhanced the degradation rate, which demonstrated that hydroxyl radical was mainly responsible for the degradation of DEP in ZVC acidic system under aerobic atmosphere condition, and the formation of hydroxyl radical was attributed to the Fenton-like reaction of in situ formed Cu(+) with H2O2.
Product Description
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Diethyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Diethyl Phthalate, Pharmaceutical Secondary Standard; Certified Reference Material
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)