Merck
  • Sex differences in liver toxicity-do female and male human primary hepatocytes react differently to toxicants in vitro?
Sex differences in liver toxicity-do female and male human primary hepatocytes react differently to toxicants in vitro?
PloS one (2015-04-08)
Milena Mennecozzi, Brigitte Landesmann, Taina Palosaari, Georgina Harris, Maurice Whelan
ABSTRACT
There is increasing amount of evidence for sex variation in drug efficiency and toxicity profiles. Women are more susceptible than men to acute liver injury from xenobiotics. In general, this is attributed to sex differences at a physiological level as well as differences in pharmacokinetics and pharmacodynamics, but neither of these can give a sufficient explanation for the diverse responses to xenobiotics. Existing data are mainly based on animal models and limited data exist on in vitro sex differences relevant to humans. To date, male and female human hepatocytes have not yet been compared in terms of their responses to hepatotoxic drugs. We investigated whether sex-specific differences in acute hepatotoxicity can be observed in vitro by comparing hepatotoxic drug effects in male and female primary human hepatocytes. Significant sex-related differences were found for certain parameters and individual drugs, showing an overall higher sensitivity of female primary hepatocytes to hepatotoxicants. Moreover, our work demonstrated that high content screening is feasible with pooled primary human hepatocytes in suspension.
Product Description
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Omeprazole, solid
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
USP
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Omeprazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Omeprazole, analytical standard