Merck
  • Neuroprotective effect of Chunghyuldan from amyloid beta oligomer induced neuroinflammation in vitro and in vivo.
Neuroprotective effect of Chunghyuldan from amyloid beta oligomer induced neuroinflammation in vitro and in vivo.
Canadian journal of physiology and pharmacology (2014-05-14)
Hyo Geun Kim, Ji-Young Kim, Wei-Wan Whang, Myung Sook Oh
ABSTRACT
Microglia-mediated inflammation is a major pathological mechanism contributing to Alzheimer's disease (AD), and has been proposed as a potential therapeutic target. Chunghyuldan (CHD; Qingxue-dan in Chinese and Daio-Orengedokuto in Japanese) possesses wide-ranging biological effects, including anti-hyperlipidemic, anti-stroke, anti-inflammatory, and antioxidant activities that could affect neurological functions. In this study, we examined the effects of CHD in in-vitro and in-vivo models of AD induced by the oligomeric form of amyloid-beta (Aβ oligomer), which acts directly on microglia-mediated neuroinflammation to result in neuronal damage and cognitive impairment. CHD at 0.1-100 μg·mL(-1) significantly protected PC12 cells and rat primary hippocampal cells from Aβ oligomer1-42 toxicity. In addition, CHD at 1-10 μg·mL(-1) inhibited Aβ oligomer1-42 induced production of nitric oxide, tumor necrosis factor-α, and interleukin-1β in microglial cells. In an in-vivo AD model, administration of CHD (50 mg·(kg body mass)(-1), for 5 days, per oral) inhibited the activation of astrocytes and microglia in the dentate gyrus and neuronal damage in the CA1 of the ipsilateral hippocampus. Moreover, CHD ameliorated cognitive impairment induced by Aβ oligomer1-42 toxicity. These results demonstrate the neuroprotective effects of CHD through inhibition of microglia-mediated neuroinflammation in in-vitro and in-vivo AD-like models induced by Aβ oligomer1-42 toxicity.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)