Merck
  • In situ formation of silver nanoparticles on thin-film composite reverse osmosis membranes for biofouling mitigation.
In situ formation of silver nanoparticles on thin-film composite reverse osmosis membranes for biofouling mitigation.
Water research (2014-06-26)
Moshe Ben-Sasson, Xinglin Lu, Edo Bar-Zeev, Katherine R Zodrow, Siamak Nejati, Genggeng Qi, Emmanuel P Giannelis, Menachem Elimelech
ABSTRACT
The potential to incorporate silver nanoparticles (Ag-NPs) as biocides in membranes for water purification has gained much interest in recent years. However, a viable strategy for loading the Ag-NPs on the membrane remains challenging. This paper presents a novel, facile procedure for loading Ag-NPs on thin-film composite (TFC) reverse osmosis membranes. Reaction of silver salt with a reducing agent on the membrane surface resulted in uniform coverage of Ag-NPs, irreversibly bound to the membrane, as confirmed by XPS, TEM, and SEM analyses. Salt selectivity of the membrane as well its surface roughness, hydrophilicity, and zeta potential were not impacted by Ag-NP functionalization, while a slight reduction (up to 17%) in water permeability was observed. The formed Ag-NPs imparted strong antibacterial activity to the membrane, leading to reduction of more than 75% in the number of live bacteria attached to the membrane for three model bacteria strains. In addition, confocal microscopy analyses revealed that Ag-NPs significantly suppressed biofilm formation, with 41% reduction in total biovolume and significant reduction in EPS, dead, and live bacteria on the functionalized membrane. The simplicity of the method, the short reaction time, the ability to load the Ag-NPs on site, and the strong imparted antibacterial activity highlight the potential of this method in real-world RO membrane applications.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%