Merck
  • Rational tailoring of substrate and inhibitor affinity via ATRP polymer-based protein engineering.
Rational tailoring of substrate and inhibitor affinity via ATRP polymer-based protein engineering.
Biomacromolecules (2014-06-24)
Hironobu Murata, Chad S Cummings, Richard R Koepsel, Alan J Russell
ABSTRACT
Atom transfer radical polymerization (ATRP)-based protein engineering of chymotrypsin with a cationic polymer was used to tune the substrate specificity and inhibitor binding. Poly(quaternary ammonium) was grown from the surface of the enzyme using ATRP after covalent attachment of a protein reactive, water-soluble ATRP-initiator. This "grafting from" conjugation approach generated a high density of cationic ammonium ions around the biocatalytic core. Modification increased the surface area of the protein over 40-fold, and the density of modification on the protein surface was approximately one chain per 4 nm(2). After modification, bioactivity was increased at low pH relative to the activity of the native enzyme. In addition, the affinity of the enzyme for a peptide substrate was increased over a wide pH range. The massively cationic chymotrypsin, which included up to 2000 additional positive charges per molecule of enzyme, was also more stable at extremes of temperature and pH. Most interestingly, we were able to rationally control the binding of two oppositely charged polypeptide protease inhibitors, aprotinin and the Bowman-Birk trypsin-chymotrypsin inhibitor from Glycine max, to the cationic derivative of chymotrypsin. This study expands upon our efforts to use polymer-based protein engineering to predictably engineer enzyme properties without the need for molecular biology.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material