Merck
  • Kinetics of sucrose conversion to fructo-oligosaccharides using enzyme (invertase) under free condition.
Kinetics of sucrose conversion to fructo-oligosaccharides using enzyme (invertase) under free condition.
Bioprocess and biosystems engineering (2014-06-19)
Deepa C Khandekar, Tapas Palai, Aman Agarwal, Prashant K Bhattacharya
ABSTRACT
The study reports the synthesis of fructo-oligosaccharide (FOS) from sucrose using invertase derived from Saccharomyces cerevisiae. The reaction was conducted in a batch mode under free enzyme condition. Fructo-oligosaccharide formation was detected at a high sucrose concentration of over 200 g/L. The investigation was extended to study the effect of different parameters such as initial sucrose concentration (ISC), pH, and enzyme concentration. A maximum FOS yield of 10 % (dry basis) was observed using 525 g/L of ISC, with 6 U/mL of the enzyme, and pH 5.5 at 40 °C. 1-Kestose was the major product of among different forms of FOS. The FOS yield increased with an increase in sucrose concentration up to 525 g/L, beyond which it started to decrease. However, the maximum FOS yield was not affected by the increasing concentration of the enzyme beyond a certain level (2 U/mL). Furthermore, the activity of enzyme slightly increased with an increase in the pH up to 6, and thereafter it declined. Addition of glucose decreased the FOS yield because of enzyme inhibition. A five-step, ten-parameter model was developed, for which the simulation was performed in COPASI. The results predicted by the model were consistent with the experimental data.
Sigma-Aldrich
4-Hydroxybenzhydrazide, ≥97%
Supelco
Nystose, analytical standard
Supelco
1-Kestose, analytical standard