Merck
  • Role of secondary metabolites in establishment of the mutualistic partnership between Xenorhabdus nematophila and the entomopathogenic nematode Steinernema carpocapsae.
Role of secondary metabolites in establishment of the mutualistic partnership between Xenorhabdus nematophila and the entomopathogenic nematode Steinernema carpocapsae.
Applied and environmental microbiology (2014-11-16)
Swati Singh, David Orr, Emmanuel Divinagracia, Joseph McGraw, Kellen Dorff, Steven Forst
ABSTRACT
Xenorhabdus nematophila engages in a mutualistic partnership with the nematode Steinernema carpocapsae, which invades insects, migrates through the gut, and penetrates into the hemocoel (body cavity). We showed previously that during invasion of Manduca sexta, the gut microbe Staphylococcus saprophyticus appeared transiently in the hemocoel, while Enterococcus faecalis proliferated as X. nematophila became dominant. X. nematophila produces diverse secondary metabolites, including the major water-soluble antimicrobial xenocoumacin. Here, we study the role of X. nematophila antimicrobials in interspecies competition under biologically relevant conditions using strains lacking either xenocoumacin (ΔxcnKL strain), xenocoumacin and the newly discovered antibiotic F (ΔxcnKL:F strain), or all ngrA-derived secondary metabolites (ngrA strain). Competition experiments were performed in Grace's insect medium, which is based on lepidopteran hemolymph. S. saprophyticus was eliminated when inoculated into growing cultures of either the ΔxcnKL strain or ΔxcnKL:F strain but grew in the presence of the ngrA strain, indicating that ngrA-derived antimicrobials, excluding xenocoumacin or antibiotic F, were required to eliminate the competitor. In contrast, S. saprophyticus was eliminated when coinjected into M. sexta with either the ΔxcnKL or ngrA strain, indicating that ngrA-derived antimicrobials were not required to eliminate the competitor in vivo. E. faecalis growth was facilitated when coinjected with either of the mutant strains. Furthermore, nematode reproduction in M. sexta naturally infected with infective juveniles colonized with the ngrA strain was markedly reduced relative to the level of reproduction when infective juveniles were colonized with the wild-type strain. These findings provide new insights into interspecies competition in a host environment and suggest that ngrA-derived compounds serve as signals for in vivo nematode reproduction.
Sigma-Aldrich
L-Arginine-7-amido-4-methylcoumarin hydrochloride, cathepsin H substrate
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard