Merck
  • Simple procedure for determination of valproic acid in dried blood spots by gas chromatography-mass spectrometry.
Simple procedure for determination of valproic acid in dried blood spots by gas chromatography-mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-04-29)
Liliane Rhoden, Marina Venzon Antunes, Paulina Hidalgo, Cleber Álvares da Silva, Rafael Linden
ABSTRACT
Valproic acid (VA) is a drug widely used to treat epilepsy and bipolar disorder, at recommended serum concentrations ranging form 50 to 100μg ml(-1). A novel option for therapeutic drug monitoring that has been emerging recently its testing using dried blood spots on paper (DBS), but there are no reports of its application to assaying VA. In this study, a methodology was developed for the determination of VA in 6mm diameter DBS, equivalent to around 12μl of blood, using gas chromatography combined with mass spectrometry. DBS were extracted with a mixture of acetonitrile and methanol (1:3, v/v). The method is linear from 5 to 250μgml(-1) with intra-assay and inter-assay precision of 2.67-8.15% and 2.28-3.67%, respectively. Accuracy was 102.84-104.42%. VA was stable in DBS stored at 45°C for up to 21 days. VA concentrations in DBS correlated with concentrations assayed in serum, with r=0.9948. Mean ratio between VA concentrations in serum and DBS in clinical samples was 1.883. Dried blood spots are a viable option for collection and transport of samples and for assaying VA in the context of therapeutic drug monitoring, especially in Developing Countries.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)