Merck
  • First characterisation of flavonoid- and diarylheptanoid-type antioxidant phenolics in Corylus maxima by HPLC-DAD-ESI-MS.
First characterisation of flavonoid- and diarylheptanoid-type antioxidant phenolics in Corylus maxima by HPLC-DAD-ESI-MS.
Journal of pharmaceutical and biomedical analysis (2015-01-17)
Eszter Riethmüller, Gergő Tóth, Ágnes Alberti, Krisztina Végh, Ilaria Burlini, Árpád Könczöl, György Tibor Balogh, Ágnes Kéry
ABSTRACT
Corylus maxima Mill. (Betulaceae) leaves have been used in traditional medicine both internally and externally, nevertheless phytochemical exploration of the plant remains incomplete. In this study, the in vitro antioxidant activity and polyphenolic composition of the ethyl acetate and methanolic extracts of C. maxima leaves and bark are reported for the first time. The radical scavenging activities of the extracts were investigated by the ABTS and DPPH assays. All the extracts of C. maxima possessed notable antioxidant activity. By mean of a HPLC-DAD-ESI-TOF and a HPLC-DAD-ESI-MS/MS method, altogether twenty-two phenolics were tentatively characterised: one flavan derivative (1), seven flavonol derivatives (4, 6, 12, 13, 16, 20 and 21) and fourteen diarylheptanoids (2, 3, 5, 7-11, 14, 15, 17-19 and 22). The amount of the two main flavonoids - myricetin-3-O-rhamnoside (6) and quercetin-3-O-rhamnoside (13) - and two diarylheptanoids - oregonin (3) and hirsutenone (15) - in the extracts were determined by a validated HPLC-ESI-MS/MS method in multiple reaction monitoring (MRM) mode. Our results showed that C. maxima could be considered as a valuable source of pharmacologically important natural products that might contribute to the revaluation of the phytotherapeutical potential of the plant.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Kaempferol, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%