Merck
  • Controlled dual delivery of BMP-2 and dexamethasone by nanoparticle-embedded electrospun nanofibers for the efficient repair of critical-sized rat calvarial defect.
Controlled dual delivery of BMP-2 and dexamethasone by nanoparticle-embedded electrospun nanofibers for the efficient repair of critical-sized rat calvarial defect.
Biomaterials (2014-12-03)
Long Li, Guangliang Zhou, Yi Wang, Guang Yang, Shan Ding, Shaobing Zhou
ABSTRACT
There is an urgent need to develop biomimetic bone tissue engineering scaffolds for the repair of critical-sized calvarial defect. In this study, we developed a new nanoparticle-embedded electrospun nanofiber scaffold for the controlled dual delivery of BMP-2 and dexamethasone (DEX). The scaffold was achieved by (1) the encapsulation of BMP-2 into bovine serum albumin (BSA) nanoparticles to maintain the bioactivity of BMP-2 and (2) the co-electrospinning of the blending solution composed of the BSA nanoparticles, DEX and the poly(ε-caprolactone)-co-poly(ethylene glycol) (PCE) copolymer. The in vitro studies showed that the bioactivity of DEX and BMP-2 was preserved in the dual-drug-loaded nanofiber scaffold, and a sequential release pattern in which most of the DEX was released in the original eight days and the BMP-2 release lasted up to 35 days was achieved. The in vitro osteogenesis study demonstrated that the drug-loaded groups exhibited a strong ability to induce differentiation toward osteoblasts. In vivo osteogenesis studies also revealed that the degrees of repair of rat calvarial defect achieved with the drug-loaded nanofiber scaffolds were significantly better than those obtained with the blank materials; in particular, the dual-drug-loaded nanofiber scaffold manifested the best repair efficacy due to a synergistic effect of BMP-2 and DEX. Therefore, the dual-drug-loaded nanofiber scaffold is deemed a strong potential candidate for the repair of bone defects in bone tissue engineering.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrachloroethylene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Supelco
Tetrachloroethylene, analytical standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Propidium iodide solution
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Density Standard 1623 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
N,O-Bis(trimethylsilyl)acetamide, derivatization grade (GC derivatization), LiChropur, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.