Merck
  • Sex differences in methamphetamine pharmacokinetics in adult rats and its transfer to pups through the placental membrane and breast milk.
Sex differences in methamphetamine pharmacokinetics in adult rats and its transfer to pups through the placental membrane and breast milk.
Drug and alcohol dependence (2014-04-15)
Lukas Rambousek, Petr Kacer, Kamila Syslova, Jakub Bumba, Vera Bubenikova-Valesova, Romana Slamberova
ABSTRACT
Methamphetamine (METH) abuse is a growing health problem worldwide, and METH use during pregnancy not only endangers the mother's health but also the developing fetus. To provide better insight into these risks, we performed the following experiments. First, we investigated how sex influences the pharmacokinetics of METH and amphetamine (AMP) in male and female rats. Subsequently, we simulated chronic exposure of prenatal infants to METH abuse by investigating brain and plasma levels of METH and AMP in dams and pups. Finally, we modeled chronic exposure of infants to METH via breast milk and investigated sex differences in pups with regard to drug levels and possible sensitization effect of chronic prenatal METH co-treatment. We observed significantly higher levels of METH and AMP in the plasma and brain of female rats compared to males. Additionally, brain concentrations of METH and AMP in pups exposed to METH prenatally were equivalent to 62.13% and 37.78% relative to dam, respectively. Plasma concentrations of AMP where equivalent to 100% of the concentration in dams, while METH was equivalent to only 36.98%. Finally, we did not observe a significant effect relative to sex with regard to METH/AMP levels or sensitization effects linked to prenatal METH exposure. We demonstrated that female rats display higher levels of METH and AMP, thus indicating a greater risk of addiction and toxicity. Furthermore, our data show that pups are exposed to both METH and AMP following dam exposure.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material