Merck
  • Implication of novel thiazolo-thiophene derivative (MCD-KV-10) for management of asthma.
Implication of novel thiazolo-thiophene derivative (MCD-KV-10) for management of asthma.
Journal of enzyme inhibition and medicinal chemistry (2014-06-19)
Dhiraj Patil, Ranjeet Prasad Dash, Sandeep Kumar Thakur, Amit N Pandya, P Venkatesh, Kamala K Vasu, Manish Nivsarkar
ABSTRACT
Asthma is multifaceted disease where many targets contribute towards its development and progression. Among these, adenosine receptor subtypes play a major role. MCD-KV-10, a novel thiazolo-thiophene was designed and evaluated pre-clinically for its implication in management of asthma. This compound showed good affinity and selectivity towards A(2A)/A3 adenosine receptor (AR) subtypes. Furthermore, MCD-KV-10 was evaluated for in vitro lipoxygenase inhibition activity; in vivo mast cell stabilization potential and in vivo anti-asthmatic activity was done in ovalbumin-induced airway inflammation model in guinea pigs. The compound showed good (>57%) inhibition of lipoxygenase enzyme and also effectively protected mast cell degranulation (>63%). The compound showed good anti-asthmatic activity as inferred from the in vivo studies. These results indicate that MCD-KV-10 has an inhibitory effect on airway inflammation. Though, we have identified a potential candidate for management of asthma, further mechanistic studies are needed.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))