Merck
  • Extracellular matrices decellularized from embryonic stem cells maintained their structure and signaling specificity.
Extracellular matrices decellularized from embryonic stem cells maintained their structure and signaling specificity.
Tissue engineering. Part A (2013-07-16)
Sébastien Sart, Teng Ma, Yan Li
ABSTRACT
Embryonic stem cells (ESCs) emerge as a promising tool for tissue engineering and regenerative medicines due to their extensive self-renewal ability and the capacity to give rise to cells from all three-germ layers. ESCs also secrete a large amount of endogenous extracellular matrices (ECMs), which play an important role in regulating ESC self-renewal, lineage commitment, and tissue morphogenesis. ECMs derived from ESCs have a broader signaling capacity compared to somatic ECMs and are predicted to have a lower risk of tumor formation associated with ESCs. In this study, ECMs from undifferentiated ESC monolayers, undifferentiated aggregates, or differentiated embryoid bodies at different developmental stages and lineage specifications were decellularized and their capacities to direct ESC proliferation and differentiation were characterized. The results demonstrate that the ESC-derived ECMs were able to influence ESC proliferation and differentiation by direct interactions with the cells and by influencing the signaling functions of the regulatory macromolecules such as retinoic acid. Such matrices have the potential to present regulatory signals to direct lineage- and development-specific cellular responses for in vitro applications or cell delivery.
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethidium bromide monoazide, ≥95% (HPLC), solid
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard