Merck
  • Differences in the transcriptional response to fulvestrant and estrogen deprivation in ER-positive breast cancer.
Differences in the transcriptional response to fulvestrant and estrogen deprivation in ER-positive breast cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-06-12)
Neill Patani, Anita K Dunbier, Helen Anderson, Zara Ghazoui, Ricardo Ribas, Elizabeth Anderson, Qiong Gao, Roger A'hern, Alan Mackay, Justin Lindemann, Robert Wellings, Jill Walker, Irene Kuter, Lesley-Ann Martin, Mitch Dowsett
ABSTRACT
Endocrine therapies include aromatase inhibitors and the selective estrogen receptor (ER) downregulator fulvestrant. This study aimed to determine whether the reported efficacy of fulvestrant over anastrozole, and high- over low-dose fulvestrant, reflect distinct transcriptional responses. Global gene expression profiles from ERα-positive breast carcinomas before and during presurgical treatment with fulvestrant (n = 22) or anastrozole (n = 81), and corresponding in vitro models, were compared. Transcripts responding differently to fulvestrant and estrogen deprivation were identified and integrated using Gene Ontology, pathway and network analyses to evaluate their potential significance. The overall transcriptional response to fulvestrant and estrogen deprivation was correlated (r = 0.61 in presurgical studies, r = 0.87 in vitro), involving downregulation of estrogen-regulated and proliferation-associated genes. The transcriptional response to fulvestrant was of greater magnitude than estrogen deprivation (slope = 0.62 in presurgical studies, slope = 0.63 in vitro). Comparative analyses identified 28 genes and 40 Gene Ontology categories affected differentially by fulvestrant. Seventeen fulvestrant-specific genes, including CAV1/2, SNAI2, and NRP1, associated with ERα, androgen receptor (AR), and TP53, in a network regulating cell cycle, death, survival, and tumor morphology. Eighteen genes responding differently to fulvestrant specifically predicted antiproliferative response to fulvestrant, but not anastrozole. Transcriptional effects of low-dose fulvestrant correlated with high-dose treatment, but were of lower magnitude (ratio = 0.29). The transcriptional response to fulvestrant has much in common with estrogen deprivation, but is stronger with distinctions potentially attributable to arrest of estrogen-independent ERα activity and involvement of AR signaling. Genes responding differently to fulvestrant may have predictive utility. These data are consistent with the clinical efficacy of fulvestrant versus anastrozole and higher dosing regimens.
Sigma-Aldrich
MISSION® esiRNA, targeting human ESR1
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Fulvestrant, European Pharmacopoeia (EP) Reference Standard