Merck
  • Polyethylene glycol modified magnetic carbon nanotubes as nanosorbents for the determination of methylprednisolone in rat plasma by high performance liquid chromatography.
Polyethylene glycol modified magnetic carbon nanotubes as nanosorbents for the determination of methylprednisolone in rat plasma by high performance liquid chromatography.
Journal of chromatography. A (2014-05-20)
Panfeng Yu, Hongwei Ma, Yong Shang, Ji Wu, Shun Shen
ABSTRACT
In this paper, polyethylene glycol modified (PEGylated) magnetic carbon nanotubes were developed as solid-phase extraction nanosorbents for the determination of methylprednisolone in rat plasma. The procedure mainly involved two steps including preparation of PEGylated magnetic nanosorbents and bioanalysis. Monodisperse magnetites (Fe3O4) anchored onto multi-walled carbon nanotubes (MWCNTs) were synthesized by a facile solvothermal synthesis method. The obtained MWCNTs-Fe3O4 nanomaterials were further non-covalently functionalized by a surfactant phospholipids-polyethylene glycol (DSPE-PEG). Owing to dispersibility and high enrichment ability, water-soluble PEGylated MWCNTs-Fe3O4 nanomaterials can provide more efficient way for the extraction of methylprednisolone than only MWCNTs-Fe3O4 used. The methylprednisolone could be easily extracted via π-π stacking interactions with PEGylated MWCNTs-Fe3O4. The captured methylprednisolone/nanosorbents were isolated from the matrix by placing a magnet, and desorbed by the elution solvent composed of acetonitrile. Extraction conditions such as amount of nanosorbents added, adsorption time, desorption solvent, and desorption time were investigated and optimized. The method recoveries were obtained from 88.2% to 92.9%. Limits of quantification and limits of detection of 0.01 and 0.005μg/mL were acquired, respectively. The precision ranged from 4.2% to 7.8% for within-day measurement, and for between-day variation was in the range of 5.5-9.0%. Moreover, the analytical performance obtained by PEGylated magnetic MWCNTs was compared with that of magnetic MWCNTs. The results indicated that the approach based on PEGylated magnetic MWCNTs was useful for the analysis of methylprednisolone in the complex plasma.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%