Merck
  • Annexin-A1 restricts Th17 cells and attenuates the severity of autoimmune disease.
Annexin-A1 restricts Th17 cells and attenuates the severity of autoimmune disease.
Journal of autoimmunity (2015-01-27)
Samia Yazid, Peter J Gardner, Livia Carvalho, Colin J Chu, Roderick J Flower, Egle Solito, Richard W J Lee, Robin R Ali, Andrew D Dick
ABSTRACT
Annexin-A1 (Anx-A1) is an endogenous anti-inflammatory molecule and while described as a repressor of innate immune responses, the role of Anx-A1 in adaptive immunity, and in particular in T helper (Th) cell responses, remains controversial. We have used a T-cell mediated mouse model of retinal autoimmune disease to unravel the role of Anx-A1 in the development of autoreactive Th cell responses and pathology. RBP1-20-immunized C57BL/6 Anx-A1(-/-) mice exhibit significantly enhanced retinal inflammation and pathology as a result of an uncontrolled proliferation and activation of Th17 cells. This is associated with a limited capacity to induce SOCS3, resulting in un-restricted phosphorylation of STAT3. RBP1-20-specific CD4(+) cells from immunized Anx-A1(-/-) animals generated high levels of Th17 cells-associated cytokines. Following disease induction, daily systemic administration of human recombinant Anx-A1 (hrAnx-A1), during the afferent phase of disease, restrained autoreactive CD4(+) cell proliferation, reduced expression of pro-inflammatory cytokines IL-17, IFN-γ and IL-6 and attenuated autoimmune retinal inflammatory disease. Furthermore, in man, Anx-A1 serum levels when measured in active uveitis patient sera were low and associated with the detection of IgM and IgG anti-Anx-A1 antibodies when compared to healthy individuals. This data supports Anx-A1 as an early and critical regulator of Th17 cell driven autoimmune diseases such as uveitis.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)