Merck
  • Effect of Cnidii Rhizoma on nitric oxide production and invasion of human colorectal adenocarcinoma HT-29 cells.
Effect of Cnidii Rhizoma on nitric oxide production and invasion of human colorectal adenocarcinoma HT-29 cells.
Oncology letters (2014-12-02)
Kyung-Soo Nam, Byung Geun Ha, Yun-Hee Shon
ABSTRACT
Colorectal adenocarcinoma is the most common type of gastrointestinal cancer. Colon adenocarcinoma is a major health problem worldwide due to the high prevalence and mortality rates associated with the disease. The majority of colorectal carcinomas are adenocarcinomas, which originate from the epithelial cells of the colorectal mucosa. HT-29 cells, which originate from human colon adenocarcinoma, are used as an
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Tetrazolium Blue Chloride, suitable for microbiology, ≥90% (T)
Sigma-Aldrich
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard