Merck
  • A comparison of chemiluminescent acridinium dimethylphenyl ester labels with different conjugation sites.
A comparison of chemiluminescent acridinium dimethylphenyl ester labels with different conjugation sites.
Organic & biomolecular chemistry (2015-01-13)
Anand Natrajan, David Wen
ABSTRACT
Chemiluminescent acridinium dimethylphenyl esters are highly sensitive labels that are used in automated assays for clinical diagnosis. Light emission from these labels and their conjugates is triggered by treatment with alkaline peroxide. Conjugation of acridinium ester labels is normally done at the phenol. During the chemiluminescent reaction of these acridinium esters, the phenolic ester is cleaved and the light emitting acridone moiety is liberated from its conjugate partner. In the current study, we report the synthesis of three new acridinium esters with conjugation sites at the acridinium nitrogen and compare their properties with that of a conventional acridinium ester with a conjugation site at the phenol. Our study is the first that provides a direct comparison of the emissive properties of acridinium dimethylphenyl esters (free labels and protein conjugates) with different conjugation sites, one where the light emitting acridone remains attached to its conjugate partner versus conventional labeling which results in cleavage of the acridone from the conjugate. Our results indicate that the conjugation at the acridinium nitrogen, which also alters how the acridinium ring and phenol are oriented with respect to the protein surface, has a minimal impact on emission kinetics and emission spectra. However, this mode of conjugation to three different proteins led to a significant increase in light yield which should be useful for improving the assay sensitivity.
Product Description
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
trans-Sinapic acid, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard