Merck
  • Development and validation of a high-performance liquid chromatography-fluorescence detection method for the accurate quantification of colistin in human plasma.
Development and validation of a high-performance liquid chromatography-fluorescence detection method for the accurate quantification of colistin in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-16)
Divyabharathi Chepyala, I-Lin Tsai, Hsin-Yun Sun, Shu-Wen Lin, Ching-Hua Kuo
ABSTRACT
Recently, colistin has become one of the most important drugs for treating infections caused by multidrug-resistant Gram-negative bacteria. Therapeutic drug monitoring is recommended to ensure the safety and efficacy of colistin and to improve clinical outcomes. This study developed an accurate and sensitive high-performance liquid chromatography-fluorescence detection (HPLC-FLD) method for the quantification of colistin in human plasma. The sample preparation included protein precipitation using trichloroacetic acid (TCA) and methanol, followed by in-solid phase extraction (In-SPE) derivatization with 9-fluorenylmethyl chloroformate (FMOC-Cl). A Poroshell 120 EC-C18 2.1×100mm (2.7μm) column was used in the HPLC method with a mobile phase composed of acetonitrile (ACN), tetrahydrofuran (THF), and deionized (DI) water (82%, 2%, 16% (v/v), respectively). Polymyxin B1 was used as the internal standard. The total analysis time was 22min under optimal separation conditions. The HPLC-FLD method was validated over a therapeutic range of 0.3-6.0μgmL(-1). The intra-day and inter-day precisions for colistin A and colistin B were below 9.9% and 4.5% relative standard deviations, respectively. The accuracy test results were between 100.2 and 118.4%. The extraction recoveries were between 81.6 and 94.1%. The method was linear over the test range, with a 0.9991 coefficient of determination. The limit of detection was 0.1μgmL(-1). The validated HPLC-FLD method was successfully applied to quantify the colistin concentrations in 2 patient samples for therapeutic drug monitoring.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Fmoc chloride, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Fmoc chloride, BioReagent, ≥99.0% (HPLC)
Supelco
Fmoc chloride, derivatization grade (HPLC), LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Daptomycin, cyclic lipopeptide antibiotic