Merck
  • Association of inflammatory response and oxidative injury in the pathogenesis of liver steatosis and insulin resistance following subchronic exposure to malathion in rats.
Association of inflammatory response and oxidative injury in the pathogenesis of liver steatosis and insulin resistance following subchronic exposure to malathion in rats.
Environmental toxicology and pharmacology (2014-09-03)
Mohamed Montassar Lasram, Ines Bini Dhouib, Kahna Bouzid, Aicha Jrad Lamine, Alya Annabi, Nadia Belhadjhmida, Malika Ben Ahmed, Saloua El Fazaa, Jaouida Abdelmoula, Najoua Gharbi
ABSTRACT
Insulin resistance and risk of type 2 diabetes are the most important complications following exposure to organophosphorous (OPs) pesticides. Regarding the importance of liver on metabolic pathways regulation, in particular blood glucose homeostasis, we focused on liver inflammation and oxidative damages in a subchronic model of toxicity by malathion. Adult male Wistar rats of body weight 200-250g were used for the study. Malathion (200mg/kg b.w./day) was administered to rats by oral intubation for 28 days. Glycemic and insulin resistance indices, markers of liver injury, markers of inflammation and oxidative stress were assessed. Malathion-treated rats showed increased glycemia, insulinemia and glycated hemoglobin level, HOMA-IR and HOMA-β indices, plasma activities of hepatocellular enzymes, lipid peroxidation index, CD3(+)/CD4(+) and CD3(+)/CD4(+) and pro-inflammatory cytokines when decreased antioxidant status in liver was noted. Most of our study indicates that malathion promotes insulin resistance, inflammation and Hepatosteatosis in subchronic model of exposure. On the basis of biochemical and molecular findings, it is concluded that insulin resistance induced by malathion occurs through oxidative stress and related pro-inflammatory markers in a way to result in a reduced function of insulin in liver cells.
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O