Merck
  • A relationship between structural and electronic order-disorder effects and optical properties in crystalline TiO2 nanomaterials.
A relationship between structural and electronic order-disorder effects and optical properties in crystalline TiO2 nanomaterials.
Dalton transactions (Cambridge, England : 2003) (2015-01-13)
E Silva Junior, F A La Porta, M S Liu, J Andrés, J A Varela, E Longo
ABSTRACT
The focus of this paper is on the analysis of the structural and electronic order-disorder effects at long, medium and short ranges of titanium dioxide (TiO2) nanoparticles synthesized by the sol-gel process followed by the microwave-assisted solvothermal (MAS) method at low temperatures and short reaction times. X-ray diffraction (XRD), Rietveld refinement, micro-Raman (MR) spectroscopy, transmission electron microscopy (TEM) and X-ray spectroscopy (EDX) were used to characterize the TiO2 nanoparticles. Optical properties were investigated by ultraviolet-visible (UV-vis) and photoluminescence (PL) measurements performed at room temperature. XRD and Rietveld refinement confirmed the presence of the anatase and brookite phases; nonetheless anatase is the major phase. The X-ray photoelectron spectroscopy (XPS) analysis revealed the presence of only Ti(4+) but the nonstoichiometry revealed that TiO2 NPs contain defects assigned to oxygen vacancies that lead to structural and electronic order-disorder effects observed by band gap narrowing and PL wide band emission. These intermediary energy levels (shallow and deep levels) created within the band gap act as acceptors/donors of electrons and recombination centers. The oxygen vacancies (VO(x), VO˙ and VO˙˙) responsible by degree of structural order-disorder are related to distortions (tilts) on the [TiO6] octahedron and changes in the bond lengths and bond angles between oxygen and titanium atoms that gave rise to new species of cluster makers such as [TiO6]', [TiO5·VO(x)], [TiO5·VO˙] and [TiO5·VO˙˙]. This structural transformation is consistent with a redistribution of electron density from highly ordered [TiO6](x) clusters which form distorted [TiO6]' as well as complex [TiO5·VO(x)], [TiO5·VO˙] and [TiO5·VO˙˙] clusters assigned to oxygen vacancies which were understood as displacements in the oxygen atoms' position in the bond lengths (Ti-O).
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Titanium(IV) isopropoxide, packaged for use in deposition systems
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Titanium(IV) isopropoxide, 97%
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Titanium(IV) isopropoxide, ≥97.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether