Merck
  • Separations of substituted benzenes and polycyclic aromatic hydrocarbons using normal- and reverse-phase high performance liquid chromatography with UiO-66 as the stationary phase.
Separations of substituted benzenes and polycyclic aromatic hydrocarbons using normal- and reverse-phase high performance liquid chromatography with UiO-66 as the stationary phase.
Journal of chromatography. A (2014-12-03)
Wei-Wei Zhao, Chao-Yan Zhang, Zeng-Guang Yan, Li-Ping Bai, Xiayan Wang, Hongliang Huang, You-Ya Zhou, Yabo Xie, Fa-Sheng Li, Jian-Rong Li
ABSTRACT
Metal-organic frameworks (MOFs) have great potential for applications in chromatography due to their highly tailorable porous structures and unique properties. In this work, the stable MOF UiO-66 was evaluated as both a normal-phase (NP-) and a reverse-phase (RP-) stationary phase in the high performance liquid chromatography (HPLC) to separate substituted benzenes (SBs) and polycyclic aromatic hydrocarbons (PAHs). It was found that the mobile phase composition has a significant effect on the HPLC separation. Baseline RP-HPLC separations of xylene isomers; naphthalene and anthracene; naphthalene and chrysene; and naphthalene, fluorene, and chrysene were achieved using MeOH/H2O ratios of 80:20, 75:25, 85:15, and 75:25, respectively, on the UiO-66 column. Similarly, baseline NP-HPLC separations of xylene isomers and ethylbenzene; ethylbenzene, styrene, o-xylene, and m-xylene; and several PAHs were also obtained on the UiO-66 column with different mobile phase compositions. The relative standard deviations (RSDs) of retention time, peak height, peak area, and half peak width for five replicate separations of the tested analytes were within the ranges 0.2-0.4%, 0.2-1.6%, 0.7-3.9%, 0.4-1.1%, respectively. We also evaluated other critical HPLC parameters, including injected sample mass, column temperature, and the thermodynamic characters of both the RP-HPLC and the NP-HPLC separation processes. It was confirmed that the separation of SBs on a UiO-66 column was an exothermic process, controlled by both enthalpy change (ΔH) and entropy change (ΔS). The reverse shape selectivity, size selectivity, stacking effect, and electrostatic force played vital roles in the separations of these analytes. To the best of our knowledge, this method is one of the very few examples of using MOFs as the stationary phase in both NP-HPLC and RP-HPLC. MOF-based stationary phases may thus be applied in the separations and analyses of SBs and PAHs in environmental samples.
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Supelco
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Supelco
Phenanthrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Melting point standard 79-81°C, analytical standard
Supelco
Hexane, analytical standard
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
Phenanthrene, 98%
Supelco
Chrysene, analytical standard
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, analytical standard
Chrysene, BCR®, certified reference material
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%