Merck
  • Migration of myeloid cells during inflammation is differentially regulated by the cell surface receptors Slamf1 and Slamf8.
Migration of myeloid cells during inflammation is differentially regulated by the cell surface receptors Slamf1 and Slamf8.
PloS one (2015-03-24)
Guoxing Wang, Boaz J van Driel, Gongxian Liao, Michael S O'Keeffe, Peter J Halibozek, Jacky Flipse, Burcu Yigit, Veronica Azcutia, Francis W Luscinskas, Ninghai Wang, Cox Terhorst
ABSTRACT
Previous studies have demonstrated that the cell surface receptor Slamf1 (CD150) is requisite for optimal NADPH-oxidase (Nox2) dependent reactive oxygen species (ROS) production by phagocytes in response to Gram- bacteria. By contrast, Slamf8 (CD353) is a negative regulator of ROS in response to Gram+ and Gram- bacteria. Employing in vivo migration after skin sensitization, induction of peritonitis, and repopulation of the small intestine demonstrates that in vivo migration of Slamf1-/- dendritic cells and macrophages is reduced, as compared to wt mice. By contrast, in vivo migration of Slamf8-/- dendritic cells, macrophages and neutrophils is accelerated. These opposing effects of Slamf1 and Slamf8 are cell-intrinsic as judged by in vitro migration in transwell chambers in response to CCL19, CCL21 or CSF-1. Importantly, inhibiting ROS production of Slamf8-/- macrophages by diphenyleneiodonium chloride blocks this in vitro migration. We conclude that Slamf1 and Slamf8 govern ROS-dependent innate immune responses of myeloid cells, thus modulating migration of these cells during inflammation in an opposing manner.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Isoflurane, European Pharmacopoeia (EP) Reference Standard