Merck
  • Analysis of amino acid neurotransmitters from rat and mouse spinal cords by liquid chromatography with fluorescence detection.
Analysis of amino acid neurotransmitters from rat and mouse spinal cords by liquid chromatography with fluorescence detection.
Journal of pharmaceutical and biomedical analysis (2015-01-18)
Nurullah Şanlı, Sarah E Tague, Craig Lunte
ABSTRACT
A RP-LC-FL detection method has been developed to identify and quantitate four amino acid neurotransmitters including glutamic acid, glycine, taurine and γ-aminobutyric acid in rat and mouse spinal cord tissue. 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde (CBQCA) was employed for the derivatization of these neurotransmitters prior to RP-LC-FL analysis. Different parameters which influenced separation and derivatization were optimized. Under optimum conditions, linearity was achieved within the concentration ranges of 0.50-50.00 μM for all analytes with correlation coefficients from 0.9912 to 0.9997. The LODs ranged from 0.03 μM to 0.06 μM. The proposed method has been successfully applied to the determination of amino acid neurotransmitters in biological samples such as rat and mouse spinal cord with satisfactory recoveries.
Product Description
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
DL-2-Aminoadipic acid, ≥99%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)