Merck
  • Analysis of 1,2-diol diesters in vernix caseosa by high-performance liquid chromatography - atmospheric pressure chemical ionization mass spectrometry.
Analysis of 1,2-diol diesters in vernix caseosa by high-performance liquid chromatography - atmospheric pressure chemical ionization mass spectrometry.
Journal of chromatography. A (2015-01-04)
Lenka Šubčíková, Michal Hoskovec, Vladimír Vrkoslav, Tereza Čmelíková, Eva Háková, Radka Míková, Pavel Coufal, Antonín Doležal, Richard Plavka, Josef Cvačka
ABSTRACT
Fatty acid diesters of long-chain 1,2-diols (1,2-DDE), or type II wax diesters, were analyzed in the vernix caseosa of a newborn girl. 1,2-DDE were isolated from the total lipid extract by the semipreparative TLC using plates coated with silica gel. Chromatographic separation of the 1,2-DDE molecular species was achieved on the non-aqueous reversed-phase HPLC with two Nova-Pak C18 columns connected in series (a total length of 45cm) and using an acetonitrile-ethyl acetate gradient. 1,2-DDE eluted from the column in the order of their equivalent chain number. The analytes were detected as ammonium adducts by an ion-trap mass spectrometer equipped with an atmospheric pressure chemical ionization source. Their structures were elucidated using tandem mass spectrometry with MS, MS(2) and MS(3) steps in a data-dependent mode. More than two thousand molecular species of 1,2-DDE were identified in 141 chromatographic peaks. The most abundant 1,2-DDE were monounsaturated lipids consisting of a C22 diol and a C18:1 fatty acid together with C16:0, C14:0 or C15:0 fatty acids. The positions of double bonds were characterized by the fragmentation of [M+C3H5N](+) formed in the ion source.
Product Description
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Rhodamine 6G, analytical standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer