Merck
  • Meta-analysis of the effects of carvedilol versus metoprolol on all-cause mortality and hospitalizations in patients with heart failure.
Meta-analysis of the effects of carvedilol versus metoprolol on all-cause mortality and hospitalizations in patients with heart failure.
The American journal of cardiology (2015-02-25)
Alexandros Briasoulis, Mohan Palla, Luis Afonso
ABSTRACT
Long-term treatment with appropriate doses of carvedilol or metoprolol is currently recommended for patients with heart failure with reduced ejection fraction (HFrEF) to decrease the risk of death, hospitalizations, and patients' symptoms. It remains unclear if the β blockers used in patients with HFrEF are equal or carvedilol is superior to metoprolol types. We performed a meta-analysis of the comparative effects of carvedilol versus metoprolol tartrate and succinate on all-cause mortality and/or hospitalization. We conducted an Embase and MEDLINE search for prospective controlled trials and cohort studies of patients with HFrEF who were received to treatment with carvedilol versus metoprolol. We identified 4 prospective controlled and 6 cohort studies with 30,943 patients who received carvedilol and 69,925 patients on metoprolol types (tartrate and succinate) with an average follow-up duration of 36.4 months. All-cause mortality was reduced in prospective studies with carvedilol versus metoprolol tartrate. Neither all-cause mortality nor hospitalizations were significantly different between carvedilol and metoprolol succinate in the cohort studies. In conclusion, in patients with HFrEF, carvedilol and metoprolol succinate have similar effects in reducing all-cause mortality.
Product Description
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carvedilol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Carvedilol for system suitability, European Pharmacopoeia (EP) Reference Standard
Carvedilol, European Pharmacopoeia (EP) Reference Standard