Merck
  • Helicobacter pylori protects oncogenically transformed cells from reactive oxygen species-mediated intercellular induction of apoptosis.
Helicobacter pylori protects oncogenically transformed cells from reactive oxygen species-mediated intercellular induction of apoptosis.
Carcinogenesis (2014-03-26)
Georg Bauer, Stefan Bereswill, Peter Aichele, Erik Glocker
ABSTRACT
Malignant transformation of gastric epithelial cells by chronic Helicobacter pylori infection is caused by several mechanisms including attraction of reactive oxygen species (ROS)-producing neutrophils and cytotoxin-associated antigen A-mediated dysplastic alterations. Here we show that H.pylori protects transformed cells from ROS-mediated intercellular induction of apoptosis. This potential control step in oncogenesis depends on the HOCl and NO/peroxynitrite (PON) signaling pathways. Helicobacter pylori-associated catalase and superoxide dismutase (SOD) efficiently cooperate in the inhibition of HOCl and the NO/PON signaling pathways. Helicobacter pylori catalase prevents HOCl synthesis through decomposition of hydrogen peroxide. Helicobacter pylori-associated SOD interferes with the crucial interactions between superoxide anions and HOCl, as well as superoxide anions and NO. The ratio of bacteria to malignant cells is critical for sufficient protection of transformed cells. Low concentrations of H.pylori more efficiently inhibited ROS-mediated destruction of transformed cells when compared with high concentrations of bacteria. Our data demonstrate the critical role of H.pylori antioxidant enzymes in the survival of transformed cells, modulating an early step of oncogenesis that is distinct from the transformation process per se.
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%