Merck
  • Radioprotection of hematopoietic progenitors by low dose amifostine prophylaxis.
Radioprotection of hematopoietic progenitors by low dose amifostine prophylaxis.
International journal of radiation biology (2014-03-07)
Thomas M Seed, Cynthia E Inal, Vijay K Singh
ABSTRACT
Amifostine is a highly efficacious cytoprotectant when administered in vivo at high doses. However, at elevated doses, drug toxicity manifests for general, non-clinical radioprotective purposes. Various strategies have been developed to avoid toxic side-effects: The simplest is reducing the dose. In terms of protecting hematopoietic tissues, where does this effective, non-toxic minimum dose lie? C3H/HEN mice were administered varying doses of amifostine (25-100 mg/kg) 30 min prior to cobalt-60 irradiation and euthanized between 4-14 days for blood and bone marrow collection and analyses. Under steady-state, amifostine had little effect on bipotential and multi-potential marrow progenitors but marginally suppressed a more primitive, lineage negative progenitor subpopulation. In irradiated animals, prophylactic drug doses greater than 50 mg/kg resulted in significant regeneration of bipotential progenitors, moderate regeneration of multipotential progenitors, but no significant and consistent regeneration of more primitive progenitors. The low amifostine dose (25 mg/kg) failed to elicit consistent and positive, radioprotective actions on any of the progenitor subtypes. Radioprotective doses for amifostine appear to lie between 25 and 50 mg/kg. Mature, lineage-restricted progenitors appear to be more responsive to the protective effects of low doses of amifostine than the more primitive, multipotential progenitors.
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
SCF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
IL-3 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor from rat, GM-CSF, recombinant, expressed in E. coli
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Stem Cell Factor from mouse, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Stem Cell Factor human, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Macrophage Colony-Stimulating Factor human, M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor from mouse, GM-CSF, from mouse, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-3 from mouse, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
IL-3 from mouse, Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture