Merck
  • Notch pathway inhibition controls myeloma bone disease in the murine MOPC315.BM model.
Notch pathway inhibition controls myeloma bone disease in the murine MOPC315.BM model.
Blood cancer journal (2014-06-14)
R Schwarzer, N Nickel, J Godau, B M Willie, G N Duda, R Schwarzer, B Cirovic, A Leutz, R Manz, B Bogen, B Dörken, F Jundt
ABSTRACT
Despite evidence that deregulated Notch signalling is a master regulator of multiple myeloma (MM) pathogenesis, its contribution to myeloma bone disease remains to be resolved. Notch promotes survival of human MM cells and triggers human osteoclast activity in vitro. Here, we show that inhibition of Notch through the γ-secretase inhibitor XII (GSI XII) induces apoptosis of murine MOPC315.BM myeloma cells with high Notch activity. GSI XII impairs murine osteoclast differentiation of receptor activator of NF-κB ligand (RANKL)-stimulated RAW264.7 cells in vitro. In the murine MOPC315.BM myeloma model GSI XII has potent anti-MM activity and reduces osteolytic lesions as evidenced by diminished myeloma-specific monoclonal immunoglobulin (Ig)-A serum levels and quantitative assessment of bone structure changes via high-resolution microcomputed tomography scans. Thus, we suggest that Notch inhibition through GSI XII controls myeloma bone disease mainly by targeting Notch in MM cells and possibly in osteoclasts in their microenvironment. We conclude that Notch inhibition is a valid therapeutic strategy in MM.
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%